2-(3-Nitrophenyl)pyrimidine
Lieferant: Apollo Scientific
OR310543-1G
APOSOR310543-1GEA
671
EUR
APOSOR310543-1G
2-(3-Nitrophenyl)pyrimidine
2-(3-Nitrophenyl)pyrimidine
Formel:
C₁₀H₇N₃O₂ Molecular Weight: 201,181 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999076 CAS-Nummer: 69491-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...