5-[(2-Chlorophenyl)carbonyl]-1,3-thiazol-2-amine

Lieferant: Apollo Scientific

OR310544-1G
APOSOR310544-1GEA 671 EUR
APOSOR310544-1G
5-[(2-Chlorophenyl)carbonyl]-1,3-thiazol-2-amine
5-[(2-Chlorophenyl)carbonyl]-1,3-thiazol-2-amine
Formel: C₁₀H₇ClN₂OS
Molecular Weight: 238,693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26393360
CAS-Nummer: 848462-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR