1-(Amino)-2-pyrollidinone p-toluenesulphonate

Lieferant: Apollo Scientific

OR310546-1G
APOSOR310546-1GEA 52.2 EUR
APOSOR310546-1G
1-(Amino)-2-pyrollidinone p-toluenesulphonate
1-(Amino)-2-pyrollidinone p-toluenesulphonate
Formel: C₁₁H₁₆N₂O₄S
Molecular Weight: 272.3207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957343
CAS-Nummer: 924898-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR