(3R,4R)-4-Methylthio-3-[(R)-1-((t-butyldimethylsilyl)oxy)ethyl]-2-azetidinone
Lieferant: Apollo Scientific
OR310547-1G
APOSOR310547-1GEA
671
EUR
APOSOR310547-1G
(3R,4R)-4-Methylthio-3-[(R)-1-((t-butyldimethylsilyl)oxy)ethyl]-2-azetidinone
(3R,4R)-4-Methylthio-3-[(R)-1-((t-butyldimethylsilyl)oxy)ethyl]-2-azetidinone
Formel:
C₁₂H₂₅NO₂SSi Molecular Weight: 275,483 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27957344 CAS-Nummer: 82864-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...