H-Nle-OBzl·TosOH

Lieferant: Apollo Scientific

Synonyme: Benzyl (2S)-2-aminohexanoate toluene-4-sulphonic acid salt

OR310549-1G
APOSOR310549-1GEA 671 EUR
APOSOR310549-1G
H-Nle-OBzl·TosOH
H-Nle-OBzl·TosOH
Formel: C₁₃H₁₉NO₂·C₇H₈O₃S
Molecular Weight: 393,5 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 63219-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR