5,5-Dimethyl-hexahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-5'-one

Lieferant: Apollo Scientific
Achtung

OR310550-1G
APOSOR310550-1GEA 1070 EUR
APOSOR310550-1G
5,5-Dimethyl-hexahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-5'-one
5,5-Dimethyl-hexahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]-5'-one
Formel: C₁₃H₂₀O₃
Molecular Weight: 224.296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957346
CAS-Nummer: 92007-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR