2-(4-Pentylcyclohexyl)acetic acid

Lieferant: Apollo Scientific
Achtung

OR310554-1G OR310554-5G OR310554-2.5G OR310554-10G
APOSOR310554-1GEA 497 EUR
APOSOR310554-1G APOSOR310554-5G APOSOR310554-2.5G APOSOR310554-10G
2-(4-Pentylcyclohexyl)acetic acid
2-(4-Pentylcyclohexyl)acetic acid
Formel: C₁₃H₂₄O₂
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00995144
CAS-Nummer: 74603-16-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR