N-Methyl-4-nitro-α-toluenesulphonamide

Lieferant: Apollo Scientific

Synonyme: N-Methyl-(4-nitro)-benzyl sulphonamide

OR310600-5G
APOSOR310600-5GEA 220 EUR
APOSOR310600-5G
N-Methyl-4-nitro-α-toluenesulphonamide
N-Methyl-4-nitro-alpha-toluenesulphonamide
Formel: C₈H₁₀N₂O₄S
Molecular Weight: 230,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00277775
CAS-Nummer: 85952-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR