2-Iodadenosin

Lieferant: Apollo Scientific

Synonyme: (2R,3R,4S,5R)-2-(6-Amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

OR310610-1G
APOSOR310610-1GEA 120 EUR
APOSOR310610-1G
2-Iodadenosin
2-Iodadenosin
Formel: C₁₀H₁₂IN₅O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01320413
CAS-Nummer: 35109-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR