5-Amino-1- benzyl-1H-pyrazole-4-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR310613-1G
APOSOR310613-1GEA 671 EUR
APOSOR310613-1G
5-Amino-1- benzyl-1H-pyrazole-4-carbonitrile
5-Amino-1- benzyl-1H-pyrazole-4-carbonitrile
Formel: C₁₁H₁₀N₄
Molecular Weight: 198,224 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00128291
CAS-Nummer: 91091-13-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR