3,5-Dichloro-4-(hydroxymethyl)phenol

Lieferant: Apollo Scientific
Achtung

OR310627-1G
APOSOR310627-1GEA 336 EUR
APOSOR310627-1G
3,5-Dichloro-4-(hydroxymethyl)phenol
3,5-Dichloro-4-(hydroxymethyl)phenol
Formel: C₇H₆Cl₂O₂
Molecular Weight: 193,027 g/mol
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797936
CAS-Nummer: 720679-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR