3,5-Dichloro-4-formylphenyl acetate

Lieferant: Apollo Scientific

OR310629-1G
APOSOR310629-1GEA 0 EUR
APOSOR310629-1G
3,5-Dichloro-4-formylphenyl acetate
3,5-Dichloro-4-formylphenyl acetate
Formel: C₉H₆Cl₂O₃
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD27980615
CAS-Nummer: 1582770-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR