3,5-Dibrom-4-picolin
Lieferant: Apollo Scientific
|
Achtung
|
OR310632-100G
APOSOR310632-100GEA
108
EUR
APOSOR310632-100G
3,5-Dibrom-4-picolin
3,5-Dibrom-4-picolin
Formel:
C₆H₅Br₂N Schmelzpunkt: 104…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128859 CAS-Nummer: 3430-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...