4,4'-Biphenyldicarbonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4'-Dicyanobiphenyl, 4,4'-Dibenzonitrile, [1,1'-Biphenyl]-4,4'-dicarbonitrile, 4,4'-Bibenzonitrile

OR310636-1G OR310636-25G OR310636-5G
APOSOR310636-1GEA 37.7 EUR
APOSOR310636-1G APOSOR310636-25G APOSOR310636-5G
4,4'-Biphenyldicarbonitril
4,4'-Biphenyldicarbonitril
Formel: C₁₄H₈N₂
Molecular Weight: 204.23 g/mol
Schmelzpunkt: 236…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013805
CAS-Nummer: 1591-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR