2,2,6,6-Tetramethylpimelic acid
Lieferant: Apollo Scientific
Synonyme:
2,2,6,6-Tetramethylheptanedioic acid
OR310638-5G
APOSOR310638-5GEA
203
EUR
APOSOR310638-5G
2,2,6,6-Tetramethylpimelic acid
2,2,6,6-Tetramethylpimelic acid
Formel:
C₁₁H₂₀O₄ Molecular Weight: 216,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800574 CAS-Nummer: 2941-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...