4-Cyanbenzamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR310641-5G
OR310641-1G
OR310641-25G
APOSOR310641-5GEA
95.7
EUR
APOSOR310641-5G
APOSOR310641-1G
APOSOR310641-25G
4-Cyanbenzamid 98%
4-Cyanbenzamid
Formel:
C₈H₆N₂O Schmelzpunkt: 225…228 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017133 CAS-Nummer: 3034-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...