Ethyl-2-cyclohexanoneacetate

Lieferant: Apollo Scientific

Synonyme: ethyl 2-(2-oxocyclohexyl)acetate, (+/-)-2-(ethoxycarbonylmethyl)cyclohexanone, (2-oxo-cyclohexyl)-acetic acid ethyl ester

OR310644-5G
APOSOR310644-5GEA 56.7 EUR
APOSOR310644-5G
Ethyl-2-cyclohexanoneacetate
Ethyl-2-cyclohexanoneacetate
Formel: C₁₀H₁₆O₃
Molecular Weight: 184,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001638
CAS-Nummer: 24731-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR