1-(2-Furyl)-1,3-butanedione

Lieferant: Apollo Scientific

Synonyme: 1-(Furan-2-yl)butane-1,3-dione

OR310646-5G
APOSOR310646-5GEA 1010 EUR
APOSOR310646-5G
1-(2-Furyl)-1,3-butanedione
1-(2-Furyl)-1,3-butanedione
Formel: C₈H₈O₃
Molecular Weight: 152,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041759
CAS-Nummer: 25790-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR