2-Naphthylphenylketon
Lieferant: Apollo Scientific
Synonyme:
2-Benzoylnaphthalene
OR310648-5G
APOSOR310648-5GEA
148
EUR
APOSOR310648-5G
2-Naphthylphenylketon
2-Naphthyl phenyl ketone
Formel:
C₁₇H₁₂O Molecular Weight: 232,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004106 CAS-Nummer: 644-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...