Pentyl syringate

Lieferant: Apollo Scientific

OR310650-1G
APOSOR310650-1GEA 336 EUR
APOSOR310650-1G
Pentyl syringate
Pentyl syringate
Formel: C₁₄H₂₀O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD27980619
CAS-Nummer: 223907-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR