6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole

Lieferant: Apollo Scientific

OR310656-1G
APOSOR310656-1GEA 0 EUR
APOSOR310656-1G
6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole
6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole
Formel: C₁₄H₁₀N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD06410799
CAS-Nummer: 43036-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR