3-(3-Methylphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione

Lieferant: Apollo Scientific

OR310664-1G
APOSOR310664-1GEA 780 EUR
APOSOR310664-1G
3-(3-Methylphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione
3-(3-Methylphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione
Formel: C₁₅H₁₈N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD22549244
CAS-Nummer: 5955-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR