8-Methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione

Lieferant: Apollo Scientific

OR310666-1G
APOSOR310666-1GEA 0 EUR
APOSOR310666-1G
8-Methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione
8-Methyl-3-phenyl-1,4,8-triazaspiro[4.5]dec-3-ene-2-thione
Formel: C₁₄H₁₇N₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD14726872
CAS-Nummer: 892299-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR