1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one

Lieferant: Apollo Scientific

OR310672-1G
APOSOR310672-1GEA 780 EUR
APOSOR310672-1G
1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one
1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one
Formel: C₁₅H₂₁N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD20528963
CAS-Nummer: 1351398-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR