[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid

Lieferant: Apollo Scientific

OR310678-1G
APOSOR310678-1GEA 780 EUR
APOSOR310678-1G
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid
Formel: C₁₁H₉N₅O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20528959
CAS-Nummer: 1338661-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR