Ethyl-3-[(4-methylpiperazin-1-yl)sulphonyl]-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific

OR310693-1G
APOSOR310693-1GEA 780 EUR
APOSOR310693-1G
Ethyl-3-[(4-methylpiperazin-1-yl)sulphonyl]-1H-pyrazole-4-carboxylate
Ethyl-3-[(4-methylpiperazin-1-yl)sulphonyl]-1H-pyrazole-4-carboxylate
Formel: C₁₁H₁₈N₄O₄S
Molecular Weight: 302,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18194691
CAS-Nummer: 1106908-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR