4-Chloro-2-(4-methylphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde

Lieferant: Apollo Scientific

OR310701-1G
APOSOR310701-1GEA 780 EUR
APOSOR310701-1G
4-Chloro-2-(4-methylphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde
4-Chloro-2-(4-methylphenyl)pyrazolo[1,5-a]pyrazine-3-carbaldehyde
Formel: C₁₄H₁₀ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD28023510
CAS-Nummer: 1610377-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR