3-Phenyl-1,4-diazaspiro[4.4]non-3-ene-2-thione
Lieferant: Apollo Scientific
OR310704-1G
APOSOR310704-1GEA
0
EUR
APOSOR310704-1G
3-Phenyl-1,4-diazaspiro[4.4]non-3-ene-2-thione
3-Phenyl-1,4-diazaspiro[4.4]non-3-ene-2-thione
Formel:
C₁₃H₁₄N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD14735619 CAS-Nummer: 5955-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...