Dimethyl-1-(2-oxopropyl)1H-1,2,3-triazole-4,5-dicarboxylate

Lieferant: Apollo Scientific

OR310710-1G
APOSOR310710-1GEA 780 EUR
APOSOR310710-1G
Dimethyl-1-(2-oxopropyl)1H-1,2,3-triazole-4,5-dicarboxylate
Dimethyl-1-(2-oxopropyl)1H-1,2,3-triazole-4,5-dicarboxylate
Formel: C₉H₁₁N₃O₅
Molecular Weight: 241,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22549283
CAS-Nummer: 1039766-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR