3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one

Lieferant: Apollo Scientific

OR310711-1G
APOSOR310711-1GEA 780 EUR
APOSOR310711-1G
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one
3-(3-Methoxybenzyl)-2-thioxo-2,3-dihydro[1]benzofuro[3,2-d]pyrimidin-4(1H)-one
Formel: C₁₈H₁₄N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD07655393
CAS-Nummer: 892300-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR