1'H-Spiro[cycloheptane-1,2'-quinazoline]-4'-thiol

Lieferant: Apollo Scientific

OR310713-1G
APOSOR310713-1GEA 780 EUR
APOSOR310713-1G
1'H-Spiro[cycloheptane-1,2'-quinazoline]-4'-thiol
1'H-Spiro[cycloheptane-1,2'-quinazoline]-4'-thiol
Formel: C₁₄H₁₈N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28023513
CAS-Nummer: 892299-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR