1'H-Spiro[cyclopentane-1,2'-quinazoline]-4'-thiol

Lieferant: Apollo Scientific

OR310714-1G
APOSOR310714-1GEA 780 EUR
APOSOR310714-1G
1'H-Spiro[cyclopentane-1,2'-quinazoline]-4'-thiol
1'H-Spiro[cyclopentane-1,2'-quinazoline]-4'-thiol
Formel: C₁₂H₁₄N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28023514
CAS-Nummer: 346458-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR