1'H-Spiro[cyclohexane-1,2'-quinazoline]-4'-thiol

Lieferant: Apollo Scientific

OR310715-1G
APOSOR310715-1GEA 780 EUR
APOSOR310715-1G
1'H-Spiro[cyclohexane-1,2'-quinazoline]-4'-thiol
1'H-Spiro[cyclohexane-1,2'-quinazoline]-4'-thiol
Formel: C₁₃H₁₆N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28023515
CAS-Nummer: 126492-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR