Dimethyl-1-(2-oxo-2-phenylethyl)-1H-1,2,3-triazole-4,5-dicarboxylate

Lieferant: Apollo Scientific

OR310716-1G
APOSOR310716-1GEA 780 EUR
APOSOR310716-1G
Dimethyl-1-(2-oxo-2-phenylethyl)-1H-1,2,3-triazole-4,5-dicarboxylate
Dimethyl-1-(2-oxo-2-phenylethyl)-1H-1,2,3-triazole-4,5-dicarboxylate
Formel: C₁₄H₁₃N₃O₅
Molecular Weight: 303,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22549280
CAS-Nummer: 869895-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR