N'-Hydroxy-1,5-dimethyl-1H-pyrrole-2-carboximidamide

Lieferant: Apollo Scientific

OR310718-1G
APOSOR310718-1GEA 780 EUR
APOSOR310718-1G
N'-Hydroxy-1,5-dimethyl-1H-pyrrole-2-carboximidamide
N'-Hydroxy-1,5-dimethyl-1H-pyrrole-2-carboximidamide
Formel: C₇H₁₁N₃O
Molecular Weight: 153.182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03756659
CAS-Nummer: 500024-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR