1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride

Lieferant: Apollo Scientific

OR310732-1G
APOSOR310732-1GEA 780 EUR
APOSOR310732-1G
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride
Formel: C₁₃H₁₉Cl₂N₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD28023520
CAS-Nummer: 1239748-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR