4-[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

Lieferant: Apollo Scientific

OR310734-1G
APOSOR310734-1GEA 780 EUR
APOSOR310734-1G
4-[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride
4-[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride
Formel: C₁₄H₁₇N₃O₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27960946
CAS-Nummer: 1706460-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR