4-[3-(2-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

Lieferant: Apollo Scientific

OR310735-1G
APOSOR310735-1GEA 780 EUR
APOSOR310735-1G
4-[3-(2-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride
4-[3-(2-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride
Formel: C₁₄H₁₈ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27960949
CAS-Nummer: 1018626-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR