3-(3-Pyridin-4-yl-1,2,4-oxadiazol-5-yl)pyridin-2(1H)-one

Lieferant: Apollo Scientific

OR310739-1G
APOSOR310739-1GEA 780 EUR
APOSOR310739-1G
3-(3-Pyridin-4-yl-1,2,4-oxadiazol-5-yl)pyridin-2(1H)-one
3-(3-Pyridin-4-yl-1,2,4-oxadiazol-5-yl)pyridin-2(1H)-one
Formel: C₁₂H₈N₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16875009
CAS-Nummer: 1239787-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR