1-(2-Aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR310742-1G
APOSOR310742-1GEA 780 EUR
APOSOR310742-1G
1-(2-Aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride
1-(2-Aminoethyl)-1H-1,2,3-triazole-4-carboxylic acid hydrochloride
Formel: C₅H₉ClN₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28023526
CAS-Nummer: 1232979-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR