{2-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride
Lieferant: Apollo Scientific
OR310744-1G
APOSOR310744-1GEA
780
EUR
APOSOR310744-1G
{2-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride
{2-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride
Formel:
C₁₀H₁₁BrClN₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD28023529 CAS-Nummer: 937666-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...