3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride

Lieferant: Apollo Scientific

OR310752-1G
APOSOR310752-1GEA 780 EUR
APOSOR310752-1G
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride
Formel: C₁₂H₂₂Cl₂N₄
Molecular Weight: 293.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25975127
CAS-Nummer: 1443423-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR