4-Aminobenzamidin Hydrochlorid

Lieferant: Apollo Scientific

Synonyme: 4-Aminophenylamidin Hydrochlorid, 4-Aminobenzoesäureamidin Hydrochlorid

OR310754-1G
APOSOR310754-1GEA 88.6 EUR
APOSOR310754-1G
4-Aminobenzamidin Hydrochlorid
4-Aminobenzamidin Hydrochlorid
Formel: C₇H₁₀ClN₃
Molecular Weight: 171,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077095
CAS-Nummer: 7761-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR