4-[(1-Oxo-7,8,9,10-tetrahydropyrazino[1,2-b]indazol-2(1H)-yl)methyl]benzoic acid

Lieferant: Apollo Scientific

OR310757-1G
APOSOR310757-1GEA 780 EUR
APOSOR310757-1G
4-[(1-Oxo-7,8,9,10-tetrahydropyrazino[1,2-b]indazol-2(1H)-yl)methyl]benzoic acid
4-[(1-Oxo-7,8,9,10-tetrahydropyrazino[1,2-b]indazol-2(1H)-yl)methyl]benzoic acid
Formel: C₁₈H₁₇N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28023534
CAS-Nummer: 1610377-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR