2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid

Lieferant: Apollo Scientific

OR310763-1G
APOSOR310763-1GEA 780 EUR
APOSOR310763-1G
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid
Formel: C₁₄H₁₀N₂O₃
Molecular Weight: 254.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20754075
CAS-Nummer: 1111000-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR