6,7-Dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde

Lieferant: Apollo Scientific

OR310766-1G
APOSOR310766-1GEA 780 EUR
APOSOR310766-1G
6,7-Dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde
6,7-Dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde
Formel: C₁₂H₁₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD07506517
CAS-Nummer: 101382-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR