3-(1H-Pyrrol-1-yl)thieno[2,3-b]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

OR310782-1G
APOSOR310782-1GEA 780 EUR
APOSOR310782-1G
3-(1H-Pyrrol-1-yl)thieno[2,3-b]pyridine-2-carboxylic acid
3-(1H-Pyrrol-1-yl)thieno[2,3-b]pyridine-2-carboxylic acid
Formel: C₁₂H₈N₂O₂S
Molecular Weight: 244,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15732197
CAS-Nummer: 1242874-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR