Methyl-4-[(acetylsulphanyl)methyl]oxane-4-carboxylate
Lieferant: Apollo Scientific
OR310783-1G
APOSOR310783-1GEA
671
EUR
APOSOR310783-1G
Methyl-4-[(acetylsulphanyl)methyl]oxane-4-carboxylate
Methyl-4-[(acetylsulphanyl)methyl]oxane-4-carboxylate
Formel:
C₁₀H₁₆O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD28023535 CAS-Nummer: 235753-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...