Bis(4-(4-propylcyclohexyl)aniline) sulphuric acid

Lieferant: Apollo Scientific

OR310784-500MG
APOSOR310784-500MGEA 560 EUR
APOSOR310784-500MG
Bis(4-(4-propylcyclohexyl)aniline) sulphuric acid
Bis(4-(4-propylcyclohexyl)aniline) sulphuric acid
Formel: C₃₀H₄₈N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD28023536
CAS-Nummer: 1980039-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR