(5,7-Dimethyl-[1,2,4]triazolo-[1,5-a]pyrimidin-2-yl)methanol

Lieferant: Apollo Scientific

Synonyme: (5,7-Dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl)methanol

OR310787-1G
APOSOR310787-1GEA 71.2 EUR
APOSOR310787-1G
(5,7-Dimethyl-[1,2,4]triazolo-[1,5-a]pyrimidin-2-yl)methanol
(5,7-Dimethyl-[1,2,4]triazolo-[1,5-a]pyrimidin-2-yl)methanol
Formel: C₈H₁₀N₄O
Molecular Weight: 178,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846959
CAS-Nummer: 54535-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR